Tetramethylrhodamine ethyl ester perchlorate (TMRE) ≥95%

Supplier: AAT BIOQUEST INC
Warning

22220
76483-570EA 122.06 USD
76483-570
Tetramethylrhodamine ethyl ester perchlorate (TMRE) ≥95%
Tetramethylrhodamine ethyl ester perchlorate (TMRE)

Positively charged rhodamine dyes (such as rhodamine esters and rosamines) are selectively localized in mitochondria, thus they are widely used for labeling mitochondria of live cells.

Formula: C₂₆H₂₇ClN₂O₇
MDL Number: MFCD00467973
CAS Number: 115532-52-0
ADR: 5,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR