3,3'-Dipropylthiadicarbocyanine iodide [DiSC3(5)] ≥95%

Supplier: AAT BIOQUEST INC

Synonyms: DiSC3(5), 3-Propyl-2-[5-[3-propyl-2(3H)-benzothiazolylidene]-1,3-pentadienyl]benzothiazolium iodide, 5-(3-Propylbenzothiazol-2-ylidene)-1-(3-propylbenzothiazolium-2-yl)-1,3-pentadiene odide, 3,3'-Dipropylthiadicarbocyanine iodide

22076
76483-544EA 124.31 USD
76483-544
3,3'-Dipropylthiadicarbocyanine iodide [DiSC3(5)] ≥95%
3,3'-Dipropylthiadicarbocyanine iodide [DiSC3(5)]

DiI, DiO, DiS, DiD and DiR dyes are a family of lipophilic fluorescent stains for labeling membranes and other hydrophobic structures.


  • Lipophilic
  • Even Cellular Staining


Caution: For Research Use Only

Formula: C₂₅H₂₇IN₂S₂
MDL Number: MFCD00075033
CAS Number: 53213-94-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR