1,1-Dioctadecyl-3,3,3,3-tetramethylindodicarbocyanine-5,5-disulfonic acid [DiIC18(5)-DS] ≥95%
Supplier: AAT Bioquest
22054
76483-534EA 124.31 USD
76483-534
1,1-Dioctadecyl-3,3,3,3-tetramethylindodicarbocyanine-5,5-disulfonic acid [DiIC18(5)-DS] ≥95%
1,1-Dioctadecyl-3,3,3,3-tetramethylindodicarbocyanine-5,5-disulfonic acid [DiIC18(5)-DS]
DiI, DiO, DiD and DiR dyes are a family of lipophilic fluorescent stains for labeling membranes and other hydrophobic structures.
  • Lipophilic
  • Even Cellular Staining
Caution: For Research Use Only