3,3'-Di-n-hexyloxacarbocyanine iodide [DiOC6(3) iodide] ≥95%

Supplier: AAT BIOQUEST INC

22046
76483-526EA 96.26 USD
76483-526
3,3'-Di-n-hexyloxacarbocyanine iodide [DiOC6(3) iodide] ≥95%
3,3'-Di-n-hexyloxacarbocyanine iodide [DiOC6(3) iodide]

DiI, DiO, DiD and DiR dyes are a family of lipophilic fluorescent stains for labeling membranes and other hydrophobic structures.


  • Lipophilic
  • Even Cellular Staining


Caution: For Research Use Only

Formula: C₂₉H₃₇IN₂O₂
MDL Number: MFCD00011954
CAS Number: 53213-82-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR