3,3-Diheptyloxacarbocyanine iodide [DiOC7(3) iodide] ≥95%
Supplier: AAT Bioquest
22040
76483-518EA 96.26 USD
76483-518
3,3-Diheptyloxacarbocyanine iodide [DiOC7(3) iodide] ≥95%
3,3-Diheptyloxacarbocyanine iodide [DiOC7(3) iodide]
DiI, DiO, DiD and DiR dyes are a family of lipophilic fluorescent stains for labeling membranes and other hydrophobic structures.
  • Lipophilic
  • Even Cellular Staining
Caution: For Research Use Only