5(6)-Carboxy-2′,7′-dichlorofluorescein diacetate N-succinimidyl ester (CDCFDA) mixture of isomers ≥90%

Supplier: AAT BIOQUEST INC

Synonyms: Carboxy-2′,7′- dichlorofluorescein diacetate, succinimidyl ester, 5 and 6 isomers, Carboxy-DCFDA-N-succinimidyl ester, CDCFDA

22026
76483-502EA 124.31 USD
76483-502
5(6)-Carboxy-2′,7′-dichlorofluorescein diacetate N-succinimidyl ester (CDCFDA) mixture of isomers ≥90%
5(6)-Carboxy-2′,7′-dichlorofluorescein diacetate N-succinimidyl ester (CDCFDA)

It is widely recognized that fluorescent labeling of cells is an effective means to determine total cell numbers or how many viable cells exist in a sample.


  • Bright Fluorescence
  • Multiple Bioconjugation Applications


Caution: For Research Use Only

Formula: C₂₉H₁₇Cl₂NO₁₁
CAS Number: 147265-60-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR