5(6)-Carboxyfluorescein diacetate mixture of isomers ≥95%

Supplier: AAT BIOQUEST INC

Synonyms: CFDA, 5(6)-Carboxy-di-O-acetylfluorescein, 5(6)-CFDA

22021
76483-496EA 96.26 USD
76483-496
5(6)-Carboxyfluorescein diacetate mixture of isomers ≥95%
5(6)-Carboxyfluorescein diacetate

It is widely recognized that fluorescent labeling of cells is an effective means to determine total cell numbers or how many viable cells exist in a sample.


  • Bright Fluorescence
  • Multiple Bioconjugation Applications


Caution: For Research Use Only

Formula: C₂₅H₁₆O₉
MDL Number: MFCD00467494
CAS Number: 124387-19-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR