Di-8-ANEPPS ≥95%

Supplier: AAT Bioquest
Warning

21497
76483-410EA 249.58 USD
76483-410
Di-8-ANEPPS ≥95%
Di-8-ANEPPS

Di-8-ANEPPS is widely used for monitoring fast membrane potential changes.

Formula: C₃₆H₅₂N₂O₃S
MDL Number: MFCD03453608
CAS Number: 157134-53-7
ADR: 5,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR