BCECF AM ester, Ultra Pure Grade calcium indicator

Supplier: AAT BIOQUEST INC
Warning

Synonyms: 2′,7′-Bis(2-carboxyethyl)-5(6′)-carboxyfluorescein acetoxymethyl ester, Spiro(isobenzofuran-1(3H),9'-(9H)xanthene)-2',7'-dipropanoic acid, BCECF AM

21203
76483-342EA 311.76 USD
76483-342
BCECF AM ester, Ultra Pure Grade calcium indicator
BCECF AM ester

Intracellular pH plays an important modulating role in many cellular events, including cell growth, calcium regulation, enzymatic activity, receptor-mediated signal transduction, ion transport, endocytosis, chemotaxis, cell adhesion and other cellular processes.


  • Excellent intracellular pH measurement
  • Ultrapure grade

Formula: C₄₂H₄₀O₂₁
MW: 808.69 g/mol
MDL Number: MFCD00036969
CAS Number: 117464-70-7
ADR: 5,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1675a34999ab3c66703d4ebc6a350e87d3d3b990&vid=1675a34999ab3c66703d4ebc6a350e87d3d3b990&prid=web&clid=SAPDAM https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=42713cce4e4173f3fad32ad83630bc0a0bc36893&vid=551a7ca767dc2cbfd9b78c943dc74e6e649c42bc&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR