BCECF AM ester calcium indicator

Supplier: AAT BIOQUEST INC
Warning

Synonyms: 2′,7′-Bis(2-carboxyethyl)-5(6′)-carboxyfluorescein acetoxymethyl ester, Spiro(isobenzofuran-1(3H),9'-(9H)xanthene)-2',7'-dipropanoic acid, BCECF AM

21202
76483-340EA 188.87 USD
76483-340
BCECF AM ester calcium indicator
BCECF AM ester

Intracellular pH plays an important modulating role in many cellular events, including cell growth, calcium regulation, enzymatic activity, receptor-mediated signal transduction, ion transport, endocytosis, chemotaxis, cell adhesion and other cellular processes.


  • Excellent intracellular pH measurement

Formula: C₄₂H₄₀O₂₁
MW: 808.69 g/mol
MDL Number: MFCD00036969
CAS Number: 117464-70-7
ADR: 5,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d117946dea4a4ad7f7757f54445886790308c9b5&vid=d117946dea4a4ad7f7757f54445886790308c9b5&prid=web&clid=SAPDAM https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=acb007751ae0978e1af43e9bb49c651c263c27ab&vid=7c919f1aa07f0e1aaef3a6f9dce62460fa335a8d&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR