Coelenterazine h ≥95%

Supplier: AAT Bioquest
Warning

21153 21159 21165
76483-316EA 96.26 USD
76483-316 76483-318 76483-320
Coelenterazine h ≥95%
Coelenterazine h
Formula: C₂₆H₂₁N₃O₂
MDL Number: MFCD00798394
CAS Number: 50909-86-9
ADR: 5,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=38ed072e7d1aaa55a91b0450b4add4bbc11c425b&vid=5eed954b5a16e534f002fe5e8208158f1df26ae2&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR