Fluo-8H™ sodium salt
Supplier: AAT Bioquest
21095
76483-246EA 249.58 USD
76483-246
Fluo-8H™ sodium salt
Fluo-8H™ sodium salt
Calcium measurements are critical for numerous biological investigations.
  • Can be loaded at rt
  • Improved calcium response
  • Significantly increased signal
  • Cell impermeant
  • Suitable for all fluorescence platforms
Caution: For research use only