Rhod-2 AM (Bromide) ≥95%, Ultra Pure Grade

Supplier: AAT BIOQUEST INC
Warning

21062 21063 21064
76483-204EA 249.58 USD
76483-204 76483-206 76483-208
Rhod-2 AM (Bromide) ≥95%, Ultra Pure Grade
Rhod-2 AM (Bromide)

Calcium measurement is critical for numerous biological investigations.

Formula: C₅₂H₅₉BrN₄O₁₉
CAS Number: 145037-81-6
ADR: 5,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d7f4d1585f24a2cb85827e15f8b165cc3b93fb2a&vid=410926bfc7570104b03e13ee875ede0887fd13aa&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR