Rhod-2 AM (Bromide) ≥90%

Supplier: AAT BIOQUEST INC
Warning

21060
76483-202EA 223.27 USD
76483-202
Rhod-2 AM (Bromide) ≥90%
Rhod-2 AM (Bromide)

Calcium measurement is critical for numerous biological investigations.

Formula: C₅₂H₅₉BrN₄O₁₉
CAS Number: 145037-81-6
ADR: 5,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR