Fura-8™ pentasodium salt
Supplier: AAT Bioquest
Warning
21058
76483-200EA 124.31 USD
76483-200
Fura-8™ pentasodium salt
Fura-8™ pentasodium salt
Although Fura-2 has become the preferred excitation-ratioable calcium indicator of choice, it has certain limitations (e.g., lower sensitivity than single wavelength calcium indicators such as Fluo-8® and Cal-520®).
  • Improved background/signal ratio
  • Excellent intracellular retention
  • Long-wavelength emission
  • Cell impermeant
Caution: For research use only
ADR: 5,II