Quin 2 potassium salt hydrate ≥95%

Supplier: AAT Bioquest
Warning

21052
76483-188EA 188.87 USD
76483-188
Quin 2 potassium salt hydrate ≥95%
Quin 2 potassium salt hydrate

Quin-2 binds calcium tightly and resembles calcium chelator EGTA in ability to bind calcium much more tightly than magnesium.

Formula: C₂₆H₂₇N₃O
MDL Number: MFCD00167201
CAS Number: 149022-19-5
ADR: 5,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR