Quin 2-Am ≥95%

Supplier: AAT BIOQUEST INC
Warning

21050
76483-186EA 223.27 USD
76483-186
Quin 2-Am ≥95%
Quin 2-Am

Quin-2 binds calcium tightly and resembles calcium chelator EGTA in ability to bind calcium much more tightly than magnesium.

Formula: C₃₈H₄₃N₃O₁₈
CAS Number: 83104-85-2
ADR: 5,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR