Fura 2-AM ≥95%
Supplier: AAT BIOQUEST INC
|
Warning
|
Synonyms:
FURA-2 pentakis(acetoxymethyl) ester, 1-[2-(5-Carboxyoxazol-2-yl)-6-aminobenzofuran-5-oxy]-2-(2'-amino-5'-methyl-phenoxy)ethane-N,N,N',N'-tetraacetic acid, pentaacetoxymethyl ester, FURA-2 acetoxymethyl ester, Fura-2 AM Ester
21020
21022
76483-148EA
124.31
USD
76483-148
76483-150
Fura 2-AM ≥95%
Fura 2-AM
Fura-2 is excitation-ratioable while Indo-1 is emission-ratioable calcium indicator.
Formula:
C₄₄H₄₇N₃O₂₄ MW: 1001.857 g/mol |
MDL Number:
MFCD00036976 CAS Number: 108964-32-5 ADR: 5 II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...