BAPTA tetrasodium salt

Supplier: AAT Bioquest
Warning

21004
76483-122EA 96.26 USD
76483-122
BAPTA tetrasodium salt
BAPTA tetrasodium salt

BAPTA (1,2-bis(o-aminophenoxy)ethane-N,N,N',N'-tetraacetic acid) is a water-soluble and cell-impermeable calcium chelator.

Formula: C₂₂H₂₄N₂O₁₀
MDL Number: MFCD00067691
CAS Number: 126824-24-6
ADR: 5,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR