Fura-8FF™ pentapotassium salt
Supplier: AAT Bioquest
Warning
20621
76483-106EA 249.58 USD
76483-106
Fura-8FF™ pentapotassium salt
Fura-8FF™ pentapotassium salt
The cell-impermeant Fura-8FF is an analog of Fura-8 with much lower calcium binding affinity, Kd ~10 µM.
  • Improved sensitivity
  • Low binding affinity
  • Long-wavelength emission
  • Cell impermeant
Caution: For research use only
ADR: 5,II