Fura-8FF™ AM
Supplier: AAT Bioquest
Warning
20620
76483-104EA 249.58 USD
76483-104
Fura-8FF™ AM
Fura-8FF™ AM
The cell-permeant Fura-8FF AM is an analog of Fura-8 AM with much lower calcium binding affinity, Kd ~10 µm.
  • Calcium-dependent fluorescence
  • Steady signal
  • Cell permeant
Caution: For research use only
ADR: 5,II