Pluronic® F-127 10% in water

Supplier: AAT BIOQUEST INC
Warning

20053
76482-990EA 96.26 USD
76482-990
Pluronic® F-127 10% in water
Poloxamer 407 (Pluronic® F-127)

Pluronic® F-127 is a nonionic surfactant that is 100% active and relatively non-toxic to cells, and frequently used with dye AM esters to improve their water solubility.


  • Extremely common surfectant
  • Multiple applications
  • Aqueous solution


Caution: Research Use Only

Formula: HO(C₂H₄O)a(C₃H₆O)b(C₂H₄O)aH
Storage Temperature: Ambient
MDL Number: MFCD00082049
CAS Number: 9003-11-6
ADR: 5,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR