Pluronic® F-127 Cell culture tested
Supplier: AAT BIOQUEST INC
20050
76482-986EA 96.26 USD
76482-986
Pluronic® F-127 Cell culture tested
Cleaning Agents Detergents and Surface Cleansers
Pluronic® F-127 is a nonionic surfactant that is 100% active and relatively non-toxic to cells, and frequently used with dye AM esters to improve their water solubility.
  • Non-toxic to cells
It is soluble at low temperature, but gels at room temperature.