Helixyte™ Green Fluorometric dsDNA Quantitation Kit *Optimized for Microplate Readers*, AAT Bioquest
Supplier: AAT BIOQUEST INC
17650
76482-944EA 249.58 USD
76482-944
Helixyte™ Green Fluorometric dsDNA Quantitation Kit *Optimized for Microplate Readers*, AAT Bioquest
Assays Nucleic Acid Assays
Helixyte™ Green dsDNA Quantitation Assay Kit can be used for selectively detecting as little as 25 pg/ml of dsDNA in the presence of ssDNA, RNA, and free nucleotides.
  • High sensitivity
  • Exceptional DNA affinity
  • Robust protocol
  • Multiple applications
Caution: For research use only.