Helixyte™ Gold Nucleic Acid Gel Stain *10000X DMSO Solution* (1 ml), AAT Bioquest
Supplier: AAT BIOQUEST INC
17595
76482-934EA 375.27 USD
76482-934
Helixyte™ Gold Nucleic Acid Gel Stain *10000X DMSO Solution* (1 ml), AAT Bioquest
Stains Electrophoresis Stains
Helixyte™ Gold is manufactured by AAT Bioquest, and it has the same chemical structure of SYBR® Gold (SYBR® is the trademark of ThermoFisher).
  • Same Spectral Properties To SYBR® Gold
  • High Sensitivity
  • Useful For Nucleic Acid Labeling In Gels
Caution: For Research Use Only