Gelite™ Orange Nucleic Acid Gel Staining Kit, AAT Bioquest
Supplier: AAT BIOQUEST INC
17594
76482-932EA 249.58 USD
76482-932
Gelite™ Orange Nucleic Acid Gel Staining Kit, AAT Bioquest
Assays Nucleic Acid Assays
Gelite™ Orange is an extremely sensitive nucleic acid gel stain for detecting DNA or RNA in gels using a standard 300 nm UV transilluminator and Polaroid 667 black-and-white print film.
  • High sensitivity
  • Exceptional DNA affinity
  • Robust protocol
  • Multiple applications
Caution: For research use only.