Gelite™ Green Nucleic Acid Gel Staining Kit, AAT Bioquest
Supplier: AAT BIOQUEST INC
17589
76482-922EA 249.58 USD
76482-922
Gelite™ Green Nucleic Acid Gel Staining Kit, AAT Bioquest
Assays Nucleic Acid Assays
Gelite™ Green is a sensitive fluorescent nucleic acid gel stain for detecting nucleic acids in agarose and polyacrylamide gels.
  • 2× more sensitive than etbr
  • Brighter signal
  • Exceptional DNA affinity
  • Robust protocol
  • Multiple applications
Caution: For research use only.