4',6'-Diamidino-2-phenylindole dihydrochloride (DAPI dihydrochloride) 10 mM in water ≥95%

Supplier: AAT BIOQUEST INC

17507
76482-846EA 96.26 USD
76482-846
4',6'-Diamidino-2-phenylindole dihydrochloride (DAPI dihydrochloride) 10 mM in water ≥95%
4',6'-Diamidino-2-phenylindole dihydrochloride (DAPI dihydrochloride)

DAPI is a fluorescent stain that binds strongly to DNA.


  • Binding-Determined Bicolor Fluorescence
  • Cell-Permeable
  • Useful For Multicolor Imaging


Caution: For Research Use Only

Formula: C₁₆H₁₅N₅·2HCl
MDL Number: MFCD00012681
CAS Number: 28718-90-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR