Actinomycin D antibiotic

Supplier: AAT BIOQUEST INC
Warning

17505
76482-844EA 96.26 USD
76482-844
Actinomycin D antibiotic
Actinomycin D

Actinomycin is a polypeptide antibiotic isolated from soil bacteria of the genus Streptomyces.


  • Well-researched
  • Traditional chemotherapy drug

Formula: C₆₂H₈₆N₁₂O₁₆
MW: 1255.5 g/mol
MDL Number: MFCD00005033
CAS Number: 50-76-0
ADR: 5,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7e0de375bc4a298f87d766e52ef5e38ab61bca38&vid=7e0de375bc4a298f87d766e52ef5e38ab61bca38&prid=web&clid=SAPDAM https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=babc4e00a60c47846c881ad19b4b58158ffacf38&vid=ba9a333f02a8b43d5f2f8d924c33b1d252eeb157&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR