Native Streptomyces avidinii Streptavidin (from Streptomyces avidinii), mFluor™ Red 780
Supplier: AAT BIOQUEST INC
16948
76482-744EA 124.31 USD
76482-744
Native Streptomyces avidinii Streptavidin (from Streptomyces avidinii), mFluor™ Red 780
Proteins and Peptides
Streptavidin conjugates are widely used together with a conjugate of biotin for specific detection of a variety of proteins, protein motifs, nucleic acids and other molecules since streptavidin has a very high binding affinity for biotin.
  • Optimized for flow cytometry
  • Useful for assay development
  • High affinity
  • Multiple applications
Caution: For research use only.