mFluor/trade; Red 700-streptavidin conjugate, Streptavidin conjugates are widely used together with a conjugate of biotin for specific detection of a variety of proteins, protein motifs, nucleic acids and other molecules since streptavidin has a very high binding affinity for biotin, Size: 100µG
Supplier: AAT Bioquest
mFluor/trade; Red 700-streptavidin conjugate, Streptavidin conjugates are widely used together with a conjugate of biotin for specific detection of a variety of proteins, protein motifs, nucleic acids and other molecules since streptavidin has a very high binding affinity for biotin, Size: 100µG