Resorufin benzyl ether ≥90%

Supplier: AAT BIOQUEST INC
Warning

15020
76482-314EA 96.26 USD
76482-314
Resorufin benzyl ether ≥90%
Resorufin benzyl ether

This fluorogenic resorufin ether is widely used for monitoring P450 activities in cell extracts and solutions.

Formula: C₁₉H₁₃NO₃
MDL Number: MFCD00171613
CAS Number: 87687-02-3
ADR: 5,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR