3-Cyano-7-ethoxycoumarin ≥95%

Supplier: AAT BIOQUEST INC
Warning

15001
76482-310EA 96.26 USD
76482-310
3-Cyano-7-ethoxycoumarin ≥95%
3-Cyano-7-ethoxycoumarin

This fluorogenic coumarin ether is widely used for monitoring P450 activities in cell extracts and solutions.

Formula: C₁₂H₉NO₃
MDL Number: MFCD00467359
CAS Number: 117620-77-6
ADR: 5,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR