Resorufin-β-D-galactopyranoside ≥95%

Supplier: AAT Bioquest
Warning

14019
76482-300EA 124.31 USD
76482-300
Resorufin-β-D-galactopyranoside ≥95%

Resorufin beta-D-galactopyranoside is a fluorogenic galactosidase substrate that generates a red fluorescent product upon interaction with the enzyme.

Formula: C₁₈H₁₇NO₈
MDL Number: MFCD00064055
CAS Number: 95079-19-9
ADR: 5,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR