FAM-cGMP PDE substrate "Green Fluorescence"
Supplier: AAT BIOQUEST INC
Warning
13604
76482-202EA 375.27 USD
76482-202
FAM-cGMP PDE substrate "Green Fluorescence"
FAM-cGMP PDE
This green fluorescent cGMP derivative is a specific substrate for phosphodiesterase (PDE) V.
  • excellent and specific fluorogenic PDE substrate
  • cGMP research tool
  • multiple applications
ADR: 5,II