Benzoyl-Arg-AMC
Supplier: AAT BIOQUEST INC
Warning
13478
76482-162EA 124.31 USD
76482-162
Benzoyl-Arg-AMC
Benzoyl-Arg-AMC
This Benzoyl-Arg-AMC is a fluorogenic aminopeptidase and endopeptidase substrate used for the detection of enzymatic activity of human alpha-thrombin.
  • alpha-thrombin detection
  • bright signal
  • multiple applications
ADR: 5,II