(Ac-WLA)2R110
Supplier: AAT BIOQUEST INC
Warning
13468
76482-144EA 124.31 USD
76482-144
(Ac-WLA)2R110
(Ac-WLA)2R110
The non-fluorescent R110 substrates generate the bright green fluorescent rhodamine 110 product that has Ex/Em = 494/521 nm, and can be easily detected with a FITC filter set.
  • protease substrate
  • bright signal
  • multiple applications
ADR: 5,II