D-(-)-Luciferin potassium salt ≥95%

Supplier: AAT BIOQUEST INC
Warning

12505 12506 12507
76481-994EA 96.26 USD
76481-994 76481-996 76481-998
D-(-)-Luciferin potassium salt ≥95%
D(-)-Luciferin potassium salt

Luciferin is the most popular and versatile bioluminescent substrate.

Formula: C₁₁H₇KN₂O₃S₂
MDL Number: MFCD00044928
CAS Number: 115144-35-9
ADR: 5,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b95864949694c304bf77dfbead0e719455e8f25b&vid=77c89f90cee052e068adbbe5db6ae96089d6dfbe&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR