Stains-all ≥95%

Supplier: AAT Bioquest
Warning

10050
76481-844EA 96.26 USD
76481-844
Stains-all ≥95%
Stains-all

Stains-all is suitable for differential staining of nucleic acids and proteins.

Formula: C₃₀H₂₇BrN₂S₂
MDL Number: MFCD00078788
CAS Number: 7423-31-6
ADR: 5,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR