hOVA antibody development
Supplier: AAT Bioquest
Warning
5606
76481-814EA 249.58 USD
76481-814
hOVA antibody development
hOVA
hOVA is the ovalbumin (OVA) modified with hexamethylenediamine to make the proteins bear more amine groups (thus more positive charges at neutral pH).
  • Protein modifier
  • Increased immunological response
ADR: 5,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=cfb91164d3bf6f9ed695eb617bf280c0f01a87b4&vid=cfb91164d3bf6f9ed695eb617bf280c0f01a87b4&prid=web&clid=SAPDAM