hBSA *Optimized for Maximum Immunization Response* (10 mg), AAT Bioquest
Supplier: AAT Bioquest
5601
76481-810EA 249.58 USD
76481-810
hBSA *Optimized for Maximum Immunization Response* (10 mg), AAT Bioquest
Sera (Serum)
hBSA is the bovine serum albumin modified with hexamethylenediamine to make the proteins bear more amine groups (thus more positive charges at neutral pH).
  • Lyophilized powder
  • Excellent quality control
  • Stronger immune response
  • 60000 molecular weight
Caution: For research use only