FMOC-Asp(5/6-TAMRA)-OH for biomolecular labeling
Supplier: AAT Bioquest
Warning
5005
76481-710EA 626.51 USD
76481-710
FMOC-Asp(5/6-TAMRA)-OH for biomolecular labeling
FMOC-Asp(5/6-TAMRA)-OH
FMOC-Asp(5/6-TAMRA)-OH is a building block for in-sequence peptide Asp labeling by TAMRA.
  • FRET building block
  • Multiple applications
ADR: 5,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=cb1981b3bd13d1f37a19dd7814e2d55dcdcb88b6&vid=cb1981b3bd13d1f37a19dd7814e2d55dcdcb88b6&prid=web&clid=SAPDAM https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ea4b248d6577b8606a0b46af6db1c45bda1f6cf3&vid=334fecc74f5cbd253edef9816227f1e7b33dd815&prid=web&clid=SAPDAM