FMOC-Asp(5-FAM)-OH for biomolecular labeling
Supplier: AAT BIOQUEST INC
Warning
5004
76481-708EA 626.51 USD
76481-708
FMOC-Asp(5-FAM)-OH for biomolecular labeling
FMOC-Asp(5-FAM)-OH
FMOC-Asp(5-FAM)-OH is a building block for in-sequence peptide Asp labeling by FAM.
  • FRET building block
  • Multiple applications
ADR: 5,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=79c6b7842763adf4b45f48865afdd9f069739e42&vid=ea852dc09cd43716f96233bf69647e2b2380e761&prid=web&clid=SAPDAM https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7d2dbede268156939a7d043945ae5e26622c7819&vid=7d2dbede268156939a7d043945ae5e26622c7819&prid=web&clid=SAPDAM