Sulfo-SMCC (3-Sulfo-N-succinimidyl-4-(N-maleimidomethyl)cyclohexane-1-carboxylate sodium salt) for peptide synthesis

Supplier: AAT Bioquest
Warning

4503
76481-688EA 249.58 USD
76481-688
Sulfo-SMCC (3-Sulfo-N-succinimidyl-4-(N-maleimidomethyl)cyclohexane-1-carboxylate sodium salt) for peptide synthesis
Sulfo-SMCC (3-Sulfo-N-succinimidyl-4-(N-maleimidomethyl)cyclohexane-1-carboxylate sodium salt)

Compared to SMCC and sulfo-SMCC, SMCC Plus has enhanced water solubility and crosslinking efficiency.


  • Improved crosslinking efficiency
  • High water solubility

Formula: C₁₆H₁₈N₂O₉S
MDL Number: MFCD00054978
CAS Number: 92921-24-9
ADR: 5,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1cb6c02ca7ae99cf85a76f5abd6392a3ebb19948&vid=6263d686c84daa1dea49175a27461afe9e197ab3&prid=web&clid=SAPDAM https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d9f3ee33c8964434ac6bbd84043491d39c826af4&vid=d9f3ee33c8964434ac6bbd84043491d39c826af4&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR