SMCC (N-Succinimidyl 4-(N-Maleimidomethyl)cyclohexanecarboxylate) ≥85%

Supplier: AAT BIOQUEST INC
Warning

4501 4502
76481-684EA 96.26 USD
76481-684 76481-686
SMCC (N-Succinimidyl 4-(N-Maleimidomethyl)cyclohexanecarboxylate) ≥85%
SMCC (N-Succinimidyl 4-(N-Maleimidomethyl)cyclohexanecarboxylate)

SMCC is a non-cleavable and membrane permeable crosslinker with a spacer arm of 8.3 Å. It contains an amine-reactive succinimidyl ester (SE) and a sulfhydryl-reactive maleimide group. NHS esters react with primary amines at pH 7 to 9 to form stable amide bonds. Maleimides react with sulfhydryl groups at pH 6.5 to 7.5 to form stable thioether bonds. The SE group of SMCC is reacted with lysine residues on the carrier protein, converting them to reactive maleimides. SMCC is a reagent that is widely used for generating stable maleimide-activated carrier proteins that can spontaneously react with sulfhydryls. Alternatively these relatively stable maleimide-activated intermediates may be lyophilized and stored for later conjugation to a hapten.

Formula: C₁₆H₁₈N₂O₆
MDL Number: MFCD00009634
CAS Number: 64987-85-5
ADR: 5,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=adc5b82fd6acc629d1c3fce25600b47d89493f4b&vid=ccf932f6b93c8f0682db613444667f519a6b133b&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR