6-Aminohexan-1-ol for protein labeling

Supplier: AAT BIOQUEST INC
Warning

4002
76481-656EA 437.45 USD
76481-656
6-Aminohexan-1-ol for protein labeling
6-Aminohexan-1-ol

6-Aminohexanol is a precursor that is widely used for preparing biotin and dye phosphoramidites.


  • High quality
  • Phosphoramidite precursor

Formula: C₆H₁₅NO
MDL Number: MFCD00008241
CAS Number: 4048-33-3
ADR: 5,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e99219c8116cd52bcdc673f41298b52da4153aca&vid=e99219c8116cd52bcdc673f41298b52da4153aca&prid=web&clid=SAPDAM https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e5b528d876759447f9d4caa3eead4e975ac95b33&vid=ccefff95aa5bcaec5bf881cc16fd08819ca47e2d&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR