6-Aminohexan-1-ol ≥95%

Supplier: AAT Bioquest
Warning

4001
76481-654EA 124.31 USD
76481-654
6-Aminohexan-1-ol ≥95%
6-Aminohexan-1-ol

6-Aminohexanol is a precursor that is widely used for preparing biotin and dye phosphoramidites.

Formula: C₆H₁₅NO
MDL Number: MFCD00008241
CAS Number: 4048-33-3
ADR: 5,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR