Biocytin hydrazide hydrochloride ≥95%

Supplier: AAT Bioquest
Warning

3086
76481-652EA 96.26 USD
76481-652
Biocytin hydrazide hydrochloride ≥95%
Biocytin hydrazide hydrochloride

This water-soluble biocytin hydrazide is used for the selective non-radioactive detection of glycoconjugates.

Formula: C₁₆H₃₁ClN₆O₃S
CAS Number: 102743-85-1
ADR: 5,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR