Biocytin C2 maleimide biodetection

Supplier: AAT Bioquest
Warning

3085
76481-650EA 249.58 USD
76481-650
Biocytin C2 maleimide biodetection
Biocytin C2 maleimide

Biocytin maleimide readily reacts with thiol moieties of biopolymers to form thioether conjugate that is quite stable.


  • Thiol-reactive
  • Multiple applications

Formula: C₂₂H₃₄N₆O₅S
CAS Number: 1248344-33-3
ADR: 5,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a57baa83316552a0c2cb9e28d915cf10d6d6a013&vid=00cccb49ecaf26157ec6835da719a85ed0dd541d&prid=web&clid=SAPDAM https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6f4dbceb441216b859177bf143a4cf217142d8ea&vid=6f4dbceb441216b859177bf143a4cf217142d8ea&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR