D-Biotin-N-succinimidyl ester, ReadiView™ biodetection

Supplier: AAT Bioquest
Warning

3059
76481-646EA 124.31 USD
76481-646
D-Biotin-N-succinimidyl ester, ReadiView™ biodetection
D-Biotin-N-succinimidyl ester

Biotin/avidin complexes are widely applied for a variety of biological detections.


  • High fluorescence enhancement
  • Sensitive detection
  • Multiple applications

Formula: C₁₄H₁₉N₃O₅S
MDL Number: MFCD00078531
CAS Number: 35013-72-0
ADR: 5,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8ee280f5fb35f49f0f1ec97b6b2e12ca8f1d52c0&vid=4011c44a9b355ad6dce785449d06ca19e4bf6e0a&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR